Order Entry
Canada
ContactUsLinkComponent
2-Propanol standard for the analysis of tobacco, Supelco®
Flammable
Irritant
2-Propanol standard for the analysis of tobacco, Supelco®
Catalog # CA1.00272.2500
Supplier:  MilliporeSigma
CAS Number:  
undefined
Flammable
Irritant
2-Propanol standard for the analysis of tobacco, Supelco®
Catalog # CA1.00272.2500
Supplier:  MilliporeSigma
Supplier Number:  1.00272.2500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. no.:
    CA1.00272.2500
  • Supplier No.:
    1.00272.2500

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -89 °C
  • Boiling Point:
    82 °C
  • Vapor Pressure:
    41.6 hPa
  • Flash Point:
    12 °C
  • Refractive Index:
    1.378
  • Explosion Limit:
    2 - 12 %(V)
  • Density:
    0.78 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    425 °C
  • Evaporation number:
    2.93
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674