Order Entry
Canada
ContactUsLinkComponent
Recombinant Taq DNA Polymerase, Cytiva
Recombinant Taq DNA Polymerase, Cytiva
Supplier:  Cytiva
Certificates
undefined
Recombinant Taq DNA Polymerase, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • Storage conditions:
    −20 °C
  • Concentration:
    5000 units/mL

Specifications

Frequently Bought Together