Exact Volume Transfer Pipettes, Globe Scientific
Supplier: Avantor
Certificates
About this item
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
- Non-toxic and chemically inert to biological fluids and most acids
- Easy to grip with gloves
- Temperature range: –50 to 90 °C
- Can be gas (EtO) sterilized
Easy-to-use exact volume transfer pipettes are economical and accurate. They are produced from chemically resistant material and are designed to dispense precise volumes. Available in 5 sizes.
Customers Who Bought This Also Bought