Wall Mounted Drying Rack, Globe Scientific
Product Details & Documents
This 72-place wall mountable laboratory drying rack is ideal for draining and drying all types of Labware
The back plate of the support is made from a single mold of 4 mm rigid high-impact polystyrene (HIPS). The peg sockets feature closed ends, eliminating leakage and consequent biohazards. The seamless drip channel collects waste which drains via a supplied drain tube.
The 72 (push in) pegs can accommodate items with a 15 mm+ neck bore. They are easily removable for cleaning or to accommodate oddly shaped items. Also included are 11 small pegs to accommodate small diameter items with a neck bore of 6mm+ (such as test tubes). This durable rack will not rust or stain.
The rack is supplied with a mounting kit to facilitate convenient hanging.
Documents
Globe Scientific Tubes Caps and Racks Laboratory Supplies Brochure
Product Literature
Specifications
- Width:19.6"
- Description:Wall Mounted Drying Rack
- Height:19.6"
Accessories
Product Information
- Catalog #AvailabilityPrice
- Extra Pegs for Drying Rack, Small Diameter, 95 x 6 mm neck bore of 6 mmExtra Pegs for Drying Rack, Small Diameter, 95 x 6 mm neck bore of 6 mm10755-178Log in for availabilityPack of 11 - $66.85