ContactUsLinkComponent
Corning® BioCoat™ Intestinal Epithelium Differentiation Environment, Corning
Corning® BioCoat™ Intestinal Epithelium Differentiation Environment, Corning
Catalog # CACB355057
Supplier:  Corning
undefined
Corning® BioCoat™ Intestinal Epithelium Differentiation Environment, Corning
Catalog # CACB355057
Supplier:  Corning
Supplier Number:  355057
Specifications
  • Description:
    Intestinal Epithelium Differentiation Environment Kit
  • Cat. no.:
    CACB355057
  • Supplier No.:
    355057
Specifications
About this item
Corning BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models.
Includes 24 Corning® BioCoat™ Fibrillar Collagen Cell Culture Inserts (1µm pore size in two 24-well plates), 100mL Intestinal Epithelium Seeding Basal Medium, 400mL Intestinal Epithelium Differentiation Medium, and 0.5mL MITO+ Serum Extender.