These disposable standard and semi-micro cuvettes are designed for use in spectrophotometers. These cuvettes are engineered for high reproducibility and offer variations within the range of ±1% absorption. This is vital when batch analysis is being undertaken.
- Polystyrene Spectrum Range: 340 to 800 nm
- UV Grade PMMA Spectrum Range: 280 to 800 nm
A line of accessories are available separately. The cuvette holder is made from polyethylene and holds 12 cuvettes in place for preparation work, storage, and protection from tipping over. In storage, racks are stackable. The caps are produced from low density polyethylene and easily snap into the square cuvettes to provide a complete seal and added protection against spillage and contamination. The disposable stirring stick is produced from polystyrene, and is 90 mm long for comfortable access to the bottom of the cuvettes.