Clinical-Grade Centrifuge Tube, 15 ml, Globe Scientific
These disposable conical-bottom centrifuge tubes have been specially designed for clinical applications.
- Available in 15 and 50 ml versions
- Easy-to-read black graduation marks in 0.5 ml increments
- White writing area for sample identification
The blue, high-density polyethylene screw caps ensure a leak-resistant and tight seal. Produced from a special grade of polypropylene, these tubes offer exceptional clarity and chemical stability.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together