Polystyrene Test Tube, 13×75, Globe Scientific
Catalog #: 60819-970
Supplier: Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Diameter ×L:13 x 75
- Description:Polystyrene Test Tube
- Supplier No.:110470
Specifications
About this item
Polystyrene tubes can withstand 1500 rpm during centrifugation.
Customers Who Bought This Also Bought