Reagent Container, Globe Scientific
Supplier: Avantor
Certificates
About this item
Containers are designed for use with Cobas Mira® and Horiba ABX Analyzers.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.