Orders LinkContactUsLinkComponent
XRN-1
XRN-1
XRN-1
Certificates
XRN-1
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
XRN-1 is highly processive 5´→3´ exoribonuclease, requiring 5´ monophosphate
It also acts on 5´ monophosphate ssDNA with greatly reduced efficiency. XRN-1 is not efficient in removing RNAs with recessed 5´P, like tRNA. It does not cleave dsDNA, ssDNA and RNA which contain di, triphosphate, OH and capped 5´ ends.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA101449-366
  • CA102855-158
  • Enzyme name:
  • XRN-1
  • XRN-1
Specifications
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 20 units
    Log in for availability
    Log in for availability
    Each
    Each - $165.00
    $165.00
    Each
       $165.00
    Minimum order quantity is 1
    Supplier Number: M0338S
    Specifications:
    More
    Specifications
    Cat. No.
    CA101449-366
    Enzyme name
    XRN-1
  • 100 units
    Log in for availability
    Log in for availability
    Each
    Each - $654.00
    $654.00
    Each
       $654.00
    Minimum order quantity is 1
    Supplier Number: M0338L
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA102855-158
    Enzyme name
    XRN-1