ContactUsLinkComponent
TRIDYE™ DNA Ladder, 1 kb Plus, New England Biolabs
TRIDYE™ DNA Ladder, 1 kb Plus, New England Biolabs
Catalog # CA101227-142
CAS Number:  
undefined
TRIDYE™ DNA Ladder, 1 kb Plus, New England Biolabs
Catalog # CA101227-142
Supplier Number:  N3270S
CAS Number:  
Specifications
  • Description:
    Tri-Dye 1 kb plus DNA ladder
  • Size:
    250 gel lanes
  • Cat. no.:
    CA101227-142
Specifications
About this item
TriDye™ 2-Log DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis
The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. This digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference points.