Terminal Transferase, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
Terminal transferase (TdT) is a template independent polymerase that catalyzes the addition of deoxynucleotides to the 3' hydroxyl terminus of DNA molecules
Protruding, recessed or blunt-ended double or single-stranded DNA molecules serve as a substrate for TdT. The 58.3 kDa enzyme does not have 5' or 3' exonuclease activity. The addition of Co2+ in the reacton makes tailing more efficient.
Specifications
- Enzyme name:Terminal Transferase