ContactUsLinkComponent
T4 DNA Ligase Reaction Buffer, New England Biolabs
T4 DNA Ligase Reaction Buffer, New England Biolabs
Catalog # CA101227-368
CAS Number:  
undefined
T4 DNA Ligase Reaction Buffer, New England Biolabs
Catalog # CA101227-368
Supplier Number:  B0202S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    T4 DNA Ligase Reaction Buffer
  • Size:
    5.0 ml (5× 1.0 ml)
  • Cat. no.:
    CA101227-368
Specifications
About this item
New England Biolabs supplies a 10X reaction buffer with all of its enzymes
At a 1X concentration this reaction buffer assures optimal activity of the enzyme.