Sulfolobus DNA Polymerase IV (Lesion Bypass Polymerase), New England Biolabs
Catalog # CA101449-320
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Size:100 units
- Enzyme name:Sulfolobus DNA Polymerase IV (Lesion Bypass Polymerase)
- Cat. no.:CA101449-320
Specifications
About this item
Sulfolobus DNA Polymerase IV is a thermostable Y-family lesion-bypass DNA Polymerase that efficiently synthesizes DNA across a variety of DNA template lesions.