ContactUsLinkComponent
pSNAPf Vector, New England Biolabs
pSNAPf Vector, New England Biolabs
Catalog # CA101641-580
CAS Number:  
undefined
pSNAPf Vector, New England Biolabs
Catalog # CA101641-580
Supplier Number:  N9183S
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    pSNAPf Vector
  • Size:
    20 µg
  • Cat. no.:
    CA101641-580
  • Vector Details:
    pSNAPf
Specifications
About this item
pSNAPf Vector is a mammalian expression plasmid intended for the cloning and stable or transient expression of SNAP-tag® protein fusions in mammalian cells
This plasmid encodes SNAPf, a SNAP-tag protein, which is expressed under control of the CMV promoter. The expression vector has an IRES (internal ribosome entry site) and a neomycin resistance gene downstream of the SNAPf for the efficient selection of stable transfectants. pSNAPf Vector contains two multiple cloning sites to allow cloning of the fusion partner as a fusion to the N- or C-terminus of the SNAPf.