PNGase F
Supplier: New England Biolabs (NEB)
Certificates
About this item
Peptide-N-Glycosidase F, also known as PNGase F, is an amidase and supplied glycerol free for optimal performance in HPLC intensive methods
- Glycerol-free
PNGase F cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid and complex oligosaccharides from N-linked glycoproteins.
Specifications
- Enzyme name:PNGase F
- Enzyme synonyms:Peptide-N-Glycosidase F
- Tested applications:Modifying enzyme