ContactUsLinkComponent
Phusion HF® Buffer Pack, New England Biolabs
Phusion HF® Buffer Pack, New England Biolabs
Catalog # CA101640-764
CAS Number:  
undefined
Phusion HF® Buffer Pack, New England Biolabs
Catalog # CA101640-764
Supplier Number:  B0518S
CAS Number:  
Specifications
  • Description:
    Phusion HF Buffer Pack
  • Cat. no.:
    CA101640-764
  • Volume:
    6.0 ml
Specifications
About this item
New England Biolabs supplies a unique 5X reaction buffer and supplemental reagents for use with Thermo Scientific® Phusion High-Fidelity DNA Polymerase and associated products, all of which are manufactured and quality controlled at New England Biolabs.