Order Entry
Canada
ContactUsLinkComponent
PH.D.™-12 Phage Display Peptide Library, New England Biolabs
PH.D.™-12 Phage Display Peptide Library, New England Biolabs
Catalog # CA101227-570
CAS Number:  
undefined
PH.D.™-12 Phage Display Peptide Library, New England Biolabs
Catalog # CA101227-570
Supplier Number:  E8111L
CAS Number:  

Specifications

  • Description:
    Ph.D.-12 Phage Display Peptide Library
  • Size:
    50 exps
  • Environmentally Preferable:
  • Cat. no.:
    CA101227-570

Specifications

About this item

The Ph.D.™-12 Phage Display Peptide Library is based on a combinatorial library of random dodecapeptides fused to a minor coat protein (pIII) of M13 phage

The displayed peptide (12-mer) is expressed at the N-terminus of pIII, i.e., the first residue of the mature protein is the first randomized position. The peptide is followed by a short spacer (Gly-Gly-Gly-Ser) and then the wild-type pIII sequence. The library consists of approximately 109 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 µl of the supplied phage.