ContactUsLinkComponent
pBR322 DNA-Msp I Digest, New England Biolabs
pBR322 DNA-Msp I Digest, New England Biolabs
Catalog # CA101228-472
CAS Number:  
undefined
pBR322 DNA-Msp I Digest, New England Biolabs
Catalog # CA101228-472
Supplier Number:  N3032S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    pBR322 DNA-Msp I Digest
  • Size:
    50 gel lanes
  • Cat. no.:
    CA101228-472
Specifications
About this item
The MspI digest of pBR322 DNA yields 26 fragments
Suitable for use as molecular weight standards for agarose gel electrophoresis. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.