Order Entry
Canada
Orders LinkContactUsLinkComponent
 

MboI

MboI, 25000 units/ml, 2500 units
Catalog # CA101228-802
 :  R0147M
MboI
Catalog # CA101228-802
CAS Number:  
MboI
MboI

MboI, 25000 units/ml, 2500 units

CAS Number:  
Price
$533.99
Log in for availability
Each
 
Product Details & Documents

An E.coli strain that carries the MboI gene from Moraxella bovis (ATCC 10900).

Specifications

Specifications

  • Concentration:
    25000 units/ml
  • Description:
    MboI
  • Size:
    2500 units
  • Cat. No.:
    CA101228-802
  • Enzyme name:
    MboI

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.