ContactUsLinkComponent
Low Range ssRNA Ladder, New England Biolabs
Low Range ssRNA Ladder, New England Biolabs
Catalog # CA101418-104
CAS Number:  
undefined
Low Range ssRNA Ladder, New England Biolabs
Catalog # CA101418-104
Supplier Number:  N0364S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Low range ssRNA ladder
  • Size:
    100 gel lanes
  • Cat. no.:
    CA101418-104
Specifications
About this item
The Low Range ssRNA Ladder is a set of 6 RNA molecules produced by in vitro transcription of a mixture of 6 linear DNA templates
The ladder sizes are: 1000, 500, 300, 150, 80 and 50 bases. The 300 base fragment is at double intensity to serve as a reference band. This ladder is suitable for use as an ssRNA size standard on denaturing polyacrylamide-urea gels, and on denaturing or native agarose gels.