LONGAMP® Taq DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
LongAmp® Taq DNA Polymerase is a unique blend of Taq and Deep VentR™ DNA Polymerases
The 3´→5´ exonuclease activity of Deep VentR DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq DNA Polymerase alone. A wide range of PCR products can be generated, up to 30 kb from lambda DNA or from human genomic DNA.
Frequently Bought Together