ContactUsLinkComponent
K. lactis Protein Expression Kit, New England Biolabs
K. lactis Protein Expression Kit, New England Biolabs
Catalog # CA101294-866
CAS Number:  
undefined
K. lactis Protein Expression Kit, New England Biolabs
Catalog # CA101294-866
Supplier Number:  E1000S
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    K.lactis Protein Expression Kit
  • Cat. no.:
    CA101294-866
Specifications
About this item
The K
The gene is cloned into the integrative pKLAC-series of vectors, and may be expressed intracellularly or secreted. The K.  lactis system offers several advantages over other yeast and bacterial expression systems. Abundant overexpression of protein is achieved through high culture densities as well as the ability to integrate multiple copies of the vector. The pKLAC-series of vectors use a strong LAC4 promoter, which has been modified to lack expression in E. coli, making this system useful for expressing toxic genes. The high transformation efficiency of the supplied K.  lactis cells makes the system suitable for methods that require large numbers of transformants, such as expression cloning using cDNA libraries. For the selection of transformants, no expensive antibiotics are required. In addition, no methanol is required in growth media. Finally, the K. lactis system can express post-translationally modified proteins, making it a useful alternative to bacterial expression systems.
lactis Protein Expression Kit provides an easy method for expressing a gene of interest in the yeast Kluyveromyces lactis.