Endonuclease VIII
Supplier: New England Biolabs (NEB)
Certificates
About this item
Endonuclease VIII from E.coli acts as both an N-glycosylase and an AP-lyase.
The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating an apurinic (AP site). The AP-lyase activity cleaves 3´ and 5´ to the AP site leaving a 5´ phosphate and a 3´ phosphate. Damaged bases recognized and removed by Endonuclease VIII include urea, 5, 6- dihydroxythymine, thymine glycol, 5-hydroxy-5- methylhydanton, uracil glycol, 6-hydroxy-5, 6-dihydrothymine and methyltartronylurea. While Endonuclease VIII is similar to Endonuclease III, Endonuclease VIII has β and δ lyase activity while Endonuclease III has β lyase activity.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together