CLIP-SURFACE® 488, New England Biolabs
Catalog #: CA101447-728
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:CLIP-Surface 488
- Size:50 nmol
- Cat. no.:CA101447-728
Specifications
About this item
CLIP-Surface™ 488 is a photostable green fluorescent substrate that can be used to label CLIP-tag™ fusion proteins on the surface of living cells or in vitro
This cell impermeable substrate (BC-488) is based on ATTO-TEC dye ATTO 488 and is suitable for standard fluorescein filter sets. It has an excitation maximum at 506 nm and emission maximum at 526 nm. This package includes 50 nmol of CLIP-Surface 488 substrate, sufficient to make 10 ml of a 5 µM CLIP-tag™ fusion protein labeling solution.
Customers Who Bought This Also Bought