BG-PEG-NH2, New England Biolabs
Catalog #: CA101447-708
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:BG-PEG-NH2
- Size:2 mg
- Cat. no.:CA101447-708
Specifications
About this item
BG-PEG-NH2 is an amine-terminated building block for the one-step synthesis of SNAP-tag substrates from NHS esters or other activated carboxyl esters on labels or surfaces
This compound allows you to make custom SNAP-tag substrates for labeling SNAP-tag fusion proteins for a wide range of applications. BG-PEG-NH2 is particularly useful for the synthesis of SNAP-tag substrates with affinity ligands or solid surfaces.
Customers Who Bought This Also Bought