Order Entry
Canada
ContactUsLinkComponent
BbvI, Restriction Enzymes, New England Biolabs
BbvI, Restriction Enzymes, New England Biolabs
Catalog # CA101228-914
CAS Number:  
undefined
BbvI, Restriction Enzymes, New England Biolabs
Catalog # CA101228-914
Supplier Number:  R0173S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Source:
  • Description:
    BbvI
  • Size:
    300 units
  • Environmentally Preferable:
  • Enzyme name:
    BbvI
  • Cat. no.:
    CA101228-914

Specifications

About this item

An E.coli strain that carries the BbvI gene from Bacillus brevis (ATCC 9999).