You searched for: Weighing Boats
Weighing boats are ideal for general-purpose weighing, evaporating, dispensing, and storing applications. Compatible with balances and dryers, these pans support precise measurements. Available in flat-bottomed circle, hexagon, and diamond shaped dishes that pack away neatly. Crimped edges give aluminum models additional sturdiness while naturally durable material models have smooth edges for minimum sample loss while pouring. Glass funnel weighing boats have a pouring neck to prevent accidental spilling when handling or transferring hazardous materials.
VWR® Disposable Hexagonal Weigh Boats
Supplier: VWR International
These polystyrene weighing boats feature smooth surfaces to enable accurate pour-out with minimal sample loss.
Expand 4 Items
VWR® Disposable Square Weighing Boats
Supplier: VWR International
These square shaped polystyrene flexible weighing boats with smooth surfaces enable accurate pour-out with minimal sample loss.
Expand 19 Items
VWR® Aluminum Weighing Dish
Supplier: VWR International
The VWR® Aluminum Weighing Dish is made from a one-piece ASTM standard (B209-10) aluminum sheet, spun to shape.
Expand 1 Items
VWR® Sterile Standard Hexagonal Weighing Boats, White
Supplier: VWR International
These hexagonal weighing boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Expand 2 Items
VWR® Paper Weigh Boats
Supplier: VWR International
A simple and cost-effective solution to reduce plastic use in the lab, VWR® Paper Weighing Boats are both Eco-IN and Eco-OUT.
Expand 6 Items
VWR® Sterile Standard Square Weighing Boats, White
Supplier: VWR International
These square weighing boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Expand 2 Items
VWR® Micro Weigh Boat
Supplier: VWR International
Ideal for microbalance weighing and/or distribution of very small amounts of sampling material.
Expand 1 Items
VWR® Disposable Aluminum Weighing and Drying Pans
Supplier: VWR International
VWR M-Series Moisture Analyzer Aluminum Sample Pans, 90mm
Expand 1 Items
VWR® Disposable Weighing Boats
Supplier: VWR International
These polystyrene, diamond-shaped flexible boats feature smooth surfaces to provide accurate pour-out with minimal sample loss.
Expand 16 Items
VWR® Pour-Boat Weighing Dishes
Supplier: VWR International
Samples are easily dispensed with the pour spout design without flexing the dish.
Expand 5 Items
VWR® Disposable Aluminum Crinkle Dishes with Tabs
Supplier: VWR International
These dishes are ideal for general-purpose weighing, dispensing, and storing applications.
Expand 6 Items
VWR® Disposable Aluminum Weighing Dishes
Supplier: VWR International
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
Expand 5 Items
VWR® Aluminum Micro Weighing Dishes
Supplier: VWR International
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
Expand 7 Items
VWR® Disposable Aluminum Weighing and Drying Pans
Supplier: VWR International
These oil-free pans are ideal for general-purpose weighing, dispensing, storing, and drying applications.
Expand 5 Items
VWR® Disposable Aluminum Smooth-Wall Weighing Dishes
Supplier: VWR International
These smooth-wall, flat-bottom dishes are ideal for general-purpose weighing, dispensing, storing, sampling, and testing applications.
Expand 2 Items
VWR® Weighing Funnels
Supplier: VWR International
Combination weighing vessel/funnel for speedy, precise weighing and transfer of samples without loss.
Expand 4 Items
Ward's® Standard Weighing Boats
Supplier: Heathrow Scientific
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Expand 3 Items
Ward's® Aluminum Weighing Boats
Supplier: Heathrow Scientific
Dishes for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
Expand 3 Items
Ward's® Pour-Boat Weighing Dishes
Supplier: Heathrow Scientific
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Expand 3 Items
Kjeldahl Weighing Boats, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
Weighing boat is suitable for the syrupy and viscose substances, pulps and powders.
Expand 2 Items
Aluminium Weigh Boats, Mettler Toledo
Supplier: Mettler Toledo
4mmD X3mmW X 13mmL.
Expand 2 Items
Weighing Dishes, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
This weighing dishes are made from aluminum which is ideal for milligram weighing.
Expand 1 Items
Filter Pan Sets, METTLER TOLEDO®
Supplier: Mettler Toledo
Filter weighing pan assemblies for use with MX and UMX balances
Expand 1 Items
Whatman™ Kjeldahl Analysis Weighing Boat, Grade 609, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman parchment weigh boats are designed so users can collect, transfer, and drop both sample and weighing boat into the acid solution during a Kjeldahl analysis without influencing analytical results.
Expand 1 Items
Weighing Boats
Supplier: Heathrow Scientific
Ideal for applications where there is concern of cross contamination and 'clean' is a necessity.
Expand 1 Items
Kjeldahl Weighing Boats, N-free, Buchi
Supplier: Buchi
Weighing boats for powder, solids or viscous fluids for reliable nitrogen and protein determination according to Kjeldahl.
Expand 1 Items
Balance Pan Liners, Aluminum, Dyn-a-Med
Supplier: Dyn-A-Med
Compatible generic aluminum moisture pans for most moisture balances.
Expand 1 Items
Aluminum Weighing Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Inexpensive, disposable aluminum foil dishes with crimped sides, flat smooth bottoms, and a finger tab handle on the rim. They may be used for routine weighing and moisture testing, as well as an embedding mold.