ContactUsLinkComponent
Acyclonucleotide Set, New England Biolabs
Acyclonucleotide Set, New England Biolabs
Catalog # CA101228-420
CAS Number:  
undefined
Acyclonucleotide Set, New England Biolabs
Catalog # CA101228-420
Supplier Number:  N0460S
CAS Number:  
Specifications
  • Description:
    Acyclonucleotide Set
  • Size:
    0.5 umol of each
  • Cat. no.:
    CA101228-420
Specifications
About this item
Acyclonucleotide Set contains four separate tubes of acyNTPs (acyATP, acyCTP, acyGTP and acyTTP).
Acyclonucleotides are supplied as a dry powder. Addition of 50 µl of distilled or de-ionized (Milli-Q®) water will result in a  final concentration of 10 mM acyNTP. Acyclonucleotides (acyNTPs) act as chain terminators and are thus useful in applications that normally employ dideoxynucleotides such as DNA sequencing and SNP detection. AcyNTPs are especially useful in applications with archaeon DNA Polymerases, more specifically with Therminator™ DNA Polymerase. Therminator DNA Polymerase is an engineered enzyme with an increased capacity to incorporate analogs with altered sugars, such as ribonucleotides, dideoxynucleotides, 2´ deoxynucleotides and especially acyclo-base analogs.