Non-Welded Telescoping Base Stands, Labconco
About this item
Non-Welded Telescoping Base Stands have telescoping legs that may be adjusted to eight height positions at installation in 1" increments to provide a working height from 30" to 37".
- Easily assembled with simple hand tools
- Casters replace the fixed feet included with Telescoping Base Stands
- Installation is required
Non-welded stands are comprised of a durable 1.75" tubular, powder-coated steel frame with back panel support
Caster Kit includes four 5" diameter, non-marking polyurethane casters with toe locks. Load rating for each caster is 250 pounds. Casters have bearings for quiet operation. Casters add 11" (28 cm) to the overall depth of the base stand.
Four fixed, leveling fee are provided. The stand includes a 9" shelf, supports loads up to 1000 pounds. Fits an appropriately sized Purifier Logic+ Biosafety Cabinet, Purifier Cell Logic+ Biosafety Cabinet or PuriCare Procedure Station.
Certifications: ADA-compliant.
Ordering information: Caster Kit is available and sold separately. Not for use with stands that support Purifier Logic Class II, Type B2 Safety Cabinets.
4 Options Available Below
Product Details & Documents
Non-Welded Telescoping Base Stands have telescoping legs that may be adjusted to eight height positions at installation in 1" increments to provide a working height from 30" to 37".
- Easily assembled with simple hand tools
- Casters replace the fixed feet included with Telescoping Base Stands
- Installation is required
Non-welded stands are comprised of a durable 1.75" tubular, powder-coated steel frame with back panel support
Caster Kit includes four 5" diameter, non-marking polyurethane casters with toe locks. Load rating for each caster is 250 pounds. Casters have bearings for quiet operation. Casters add 11" (28 cm) to the overall depth of the base stand.
Four fixed, leveling fee are provided. The stand includes a 9" shelf, supports loads up to 1000 pounds. Fits an appropriately sized Purifier Logic+ Biosafety Cabinet, Purifier Cell Logic+ Biosafety Cabinet or PuriCare Procedure Station.
Certifications: ADA-compliant.
Ordering information: Caster Kit is available and sold separately. Not for use with stands that support Purifier Logic Class II, Type B2 Safety Cabinets.
Product Family Options
Product Information
- WidthFor use withAvailabilityPrice
- 89422-850Non-Welded Telescoping Base StandNon-Welded Telescoping Base Stand3 ft.3' Purifier Logic+ Biosafety Cabinet, 3' Purifier Cell Logic+ Biosafety Cabinet or 3' PuriCare Procedure StationLog in for availabilityEach$1,500.00Supplier Number: 3401003
- 89422-852Non-Welded Telescoping Base StandNon-Welded Telescoping Base Stand4 ft.4' Purifier Logic+ Biosafety Cabinet, 4' Purifier Cell Logic+ Biosafety Cabinet or 4' PuriCare Procedure StationLog in for availabilityEach$1,590.00Supplier Number: 3401004
- 89422-854Non-Welded Telescoping Base StandNon-Welded Telescoping Base Stand5 ft.5' Purifier Logic+ Biosafety Cabinet, 5' Purifier Cell Logic+ Biosafety Cabinet or 5' PuriCare Procedure StationLog in for availabilityEach$1,680.00Supplier Number: 3401005
- 89422-856Non-Welded Telescoping Base StandNon-Welded Telescoping Base Stand6 ft.6' Purifier Logic+ Biosafety Cabinet, 6' Purifier Cell Logic+ Biosafety Cabinet or 6' PuriCare Procedure StationLog in for availabilityEach$1,760.00Supplier Number: 3401006
- 89203-038Caster Kit for converting Telescoping Base Stands with Fixed Feet to Mobile StandsCaster Kit for converting Telescoping Base Stands with Fixed Feet to Mobile StandsDiscontinued-Supplier Number: -