ContactUsLinkComponent
Twin Adapter, with Connecting Hose, Glassco
  89426-432
 :  Glassco
undefined
Twin Adapter, with Connecting Hose, Glassco
  89426-432
 :  Glassco
 :  046.202.04
  • Description:
    Twin Adapter, with Connecting Hose
  • Environmentally Preferable:
  • Cat. no.:
    89426-432
  • ST Joint Size:
    24/40
  • Supplier No.:
    046.202.04
Adapter features two hose connections 180° from one another.
  • Two hose connections
  • 180° between connections
The two hose connections 180° from one another permit a flow of inert gas over the apparatus.