Blank Test Strips Without Reagents Reflectoquant®, MilliporeSigma
Catalog # CA1.16730.0001
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Blank Test Strips
- Cat. no.:CA1.16730.0001
- Supplier No.:1.16730.0001
- No. of Tests:50 strips
Specifications
About this item
When analyzing coloured samples using the Reflectoquant® System the intrinsic colouration can, in given circumstances, produce more or less pronounced false-high results.
Therefore the blank strips can be used to correct these measurement values without having to take time-consuming sample-pretreatment measures.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.