ContactUsLinkComponent
Calcium Test Method: Reflectometric With Test Strips 5 to 125 mg/L Ca Reflectoquant®, MilliporeSigma
  CA1.16125.0001
 :  
Calcium Test Method: Reflectometric With Test Strips 5 to 125 mg/L Ca Reflectoquant®, MilliporeSigma
  CA1.16125.0001
 :  1.16125.0001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Chemical method:
    Phthalein complexone
  • Description:
    Calcium Test
  • Cat. no.:
    CA1.16125.0001
  • Range:
    5 - 125 mg/L Ca
  • Supplier No.:
    1.16125.0001
  • No. of Tests:
    50 strips
Calcium is an important parameter for boiling and cooling water.
If the water contains calcium, deposits can be found in the plants and this can influence the function of the machines. Calcium also needs to be analyzed in drinking water and especially in beer.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.