ContactUsLinkComponent
Ascorbic Acid Test, 25 - 450 mg/L, Reflectoquant®, MilliporeSigma
  CA1.16981.0001
 :  
Ascorbic Acid Test, 25 - 450 mg/L, Reflectoquant®, MilliporeSigma
  CA1.16981.0001
 :  1.16981.0001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Chemical method:
    Phosphoromolybdenum blue
  • Description:
    Ascorbic Acid Test
  • Cat. no.:
    CA1.16981.0001
  • Range:
    25 - 450 mg/L C₆H₈O₆
  • Supplier No.:
    1.16981.0001
  • No. of Tests:
    50 strips
Ascorbic Acid (vitamin C) counts as an essential quality characteristic in many foods. Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
MilliporeSigma # 1.16981.0001 replaces and is identical to MilliporeSigma # 16981-1
The content of ascorbic acid is measured during the production and filling procedures in the fruit- and vegetable-processing sector as a measure to guarantee compliance of the minimum required values for preservation.