Copper Test Method: Colorimetric With Test Strips 10 - 30 - 100 - 300 mg/l Cu MQuant™, MilliporeSigma
Catalog # CA1.10003.0001
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:2,2'-Cuproin
- Description:Test with Strips
- Cat. no.:CA1.10003.0001
- Range:10-30-100-300 mg/l Cu
- Supplier No.:1.10003.0001
- No. of Tests:100 strips
Specifications
About this item
The test is suitable, for instance, for monitoring and testing of wastewater, from electroplating and copper etching process as well as waste water used in the manufacture of printing plates and printed circuits
MilliporeSigma # 1.10003.0001 replaces and is identical to MilliporeSigma # 10003-1
The test strips offer a very simple means of determining iron in all aqueous media and on metal surfaces.