Sulfite Test Method: Reflectometric With Test Strips 10 - 200 mg/L SO32- Reflectoquant®, MilliporeSigma
Catalog # CA1.16987.0001
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:Nitroprusside / Zn-hexacyanoferrate
- Description:Test with Strips
- Cat. no.:CA1.16987.0001
- Range:10 - 200 mg/l SO₃
- Supplier No.:1.16987.0001
- No. of Tests:50 strips
Specifications
About this item
Test strips for the determination of sulfite with reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Test kits with reagents for reliable documented quantitative results
- Barcode calibration
- Low analysis cost
- Test strips with minute amount of chemicals for easy disposal
The test can be used for the determination of sulfite ions, which form a red compound in reaction with the reagents, in e.g. wastewater, boiler water and boiler feed water and food after appropriate sample pretreatment.