ContactUsLinkComponent
Nitrate Test Method: Reflectometric With Test Strips 5 - 225 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
  CA1.16971.0001
 :  
Nitrate Test Method: Reflectometric With Test Strips 5 - 225 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
  CA1.16971.0001
 :  1.16971.0001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Chemical method:
    Griess' reaction
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.16971.0001
  • Range:
    5 - 225 mg/l NO₃
  • Supplier No.:
    1.16971.0001
  • No. of Tests:
    50 strips
The rapid determination of the nitrate content is of particular interest with fruit and vegetables to help decide whether a given delivery batch can be released for use in a specific product (e.g baby foods).
MilliporeSigma # 1.16971.0001 replaces and is identical to MilliporeSigma # 16971-1
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.