Peracetic Acid Test Method: Colorimetric With Test Strips 500 - 1000 - 1500 - 2000 mg/L MQuant™, MilliporeSigma
Catalog # CA1.17922.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Chemical method:Redox reaction
- Description:Test with Strips
- Cat. no.:CA1.17922.0001
- Range:500-1000-1500-2000 mg/l CH₃CO₃H
- Supplier No.:1.17922.0001
- No. of Tests:100 strips
Specifications
About this item
The peracetic acid tests for higher concentrations of peracetic acid are eminently suitable for checking that the prescribed end concentration of peracetic acid-based detergents are being maintained.