ContactUsLinkComponent
Peracetic Acid Test Method: Colorimetric With Test Strips 100 - 150 - 200 - 250 - 300 - 400 - 500 mg/L MQuant™, MilliporeSigma
Peracetic Acid Test Method: Colorimetric With Test Strips 100 - 150 - 200 - 250 - 300 - 400 - 500 mg/L MQuant™, MilliporeSigma
Catalog # CA1.10001.0001
Supplier:  MilliporeSigma
CAS Number:  
Peracetic Acid Test Method: Colorimetric With Test Strips 100 - 150 - 200 - 250 - 300 - 400 - 500 mg/L MQuant™, MilliporeSigma
Catalog # CA1.10001.0001
Supplier:  MilliporeSigma
Supplier Number:  1.10001.0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Chemical method:
    Redox reaction
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.10001.0001
  • Range:
    100-150-200-250-300-400-500 mg/l CH₃CO₃H
  • Supplier No.:
    1.10001.0001
  • No. of Tests:
    100 strips
Specifications
About this item
The peracetic acid tests for higher concentrations of peracetic acid are eminently suitable for checking that the prescribed end concentration of peracetic acid-based detergents are being maintained.
MilliporeSigma # 1.10001.0001 replaces and is identical to MilliporeSigma # 10001-1