ContactUsLinkComponent
Nickel Test Method: Colorimetric with Test Strips 10 - 25 - 100 - 250 - 500 mg/L Ni MQuant®, MilliporeSigma
Nickel Test Method: Colorimetric with Test Strips 10 - 25 - 100 - 250 - 500 mg/L Ni MQuant®, MilliporeSigma
Catalog # CA1.10006.0001
Supplier:  MilliporeSigma
CAS Number:  
Nickel Test Method: Colorimetric with Test Strips 10 - 25 - 100 - 250 - 500 mg/L Ni MQuant®, MilliporeSigma
Catalog # CA1.10006.0001
Supplier:  MilliporeSigma
Supplier Number:  1.10006.0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Chemical method:
    Diacetyl dioxime
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.10006.0001
  • Range:
    10-25-100-250-500 mg/l Ni
  • Supplier No.:
    1.10006.0001
  • No. of Tests:
    100 strips
Specifications
About this item
The nickel test can be used for the testing of waste water and electroplating solutions, in the glass and ceramic industry in the testing of catalysts and of mordants used for textile printing
MilliporeSigma # 1.10006.0001 replaces and is identical to MilliporeSigma # 10006-1
The test strips offer a very simple means of determining nickel in all aqueous media and on metal surfaces. They can thus be used to distinguish between nickelous and non nickelous metals.