ContactUsLinkComponent
Chlorine Test Method: Colorimetric With Test Strips 0 - 0.5 - 1 - 2 - 5 - 10 - 20 mg/L Cl₂ MQuant™, MilliporeSigma
  CA1.17925.0001
 :  
Chlorine Test Method: Colorimetric With Test Strips 0 - 0.5 - 1 - 2 - 5 - 10 - 20 mg/L Cl₂ MQuant™, MilliporeSigma
  CA1.17925.0001
 :  1.17925.0001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Chemical method:
    Redox reaction
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.17925.0001
  • Range:
    0,5-1-2-5-10-20 mg/l Cl₂
  • Supplier No.:
    1.17925.0001
  • No. of Tests:
    75 strips
The chlorine test is a rapid exploratory test for chlorine and is intended primarily for checking the use of chlorinated disinfectants
MilliporeSigma # 1.17925.0001 replaces and is identical to MilliporeSigma # 17925-1
Chlorinated disinfectants are still in common use all over the world, their major applications being in the chlorination of drinking water and swimming pools.