ContactUsLinkComponent
HiTrap Columns, Capto Q ImpRes, Cytiva
HiTrap Columns, Capto Q ImpRes, Cytiva
Supplier:  Cytiva
Certificates
undefined
HiTrap Columns, Capto Q ImpRes, Cytiva
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • CA97067-782
  • CA97067-780
  • Max. pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol