HiTrap Columns, Capto IEX Selection Kit, Cytiva
CA97068-042
: Cytiva
- Flow rate:< 4 mL/min
- Max. pressure:5 bar [0.5 MPa] (70 psi)
- Capacity:1 ml
- Storage:4 to 30 °C, 20% Ethanol (Q, DEAE), 20% Ethanol, 0.2 M Sodium Acetate (S)
- Column Hardware:PP
- Bed Dimensions:7×25 mm
- Cat. no.:CA97068-042
- Supplier No.:28934388
HiTrap Capto IEX Selection Kit is designed for screening the optimal Capto ion exchange or Cytiva multimodal chromatography resin prior to scale-up.
- Fast and easy selection. Five prepacked HiTrap columns, each containing a different IEX resin, including multimodal ion exchangers
- Convenient way to screen for the optimal Cytiva IEX resin for a given application
- Excellent for optimization studies, buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up
- HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
This kit contains a variety of columns prepacked with anion and cation exchange resins, allowing users to screen for the most appropriate column prior to scale-up.
: Kit includes five 1 ml HiTrap™ columns prepacked with Capto™ Q (strong anion), Capto™ S (strong cation), Capto™ DEAE (weak anion), Capto™ MMC (multimodal cation), Capto™ adhere (multimodal strong anion), as well as connectors and instructions.