Order Entry
Canada
Orders LinkContactUsLinkComponent
HiScreen Capto MMC Columns

HiScreen Capto MMC Columns

HiScreen™ Capto™ MMC, 75 µm, 7.7×100 mm, 4.7 ml
Catalog # CA97067-800
Supplier:   Cytiva
Supplier #:  28926980
HiScreen Capto MMC Columns
Catalog # CA97067-800
Supplier:  Cytiva
HiScreen Capto MMC Columns
HiScreen Capto MMC Columns

HiScreen™ Capto™ MMC, 75 µm, 7.7×100 mm, 4.7 ml

Price
$472.11
Log in for availability
Each
Product Details & Documents

HiScreen Capto MMC columns are prepacked with Capto MMC ion exchange chromatography medium, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same resin using the same linear fluid velocity

Capto MMC is a multimodal cation exchanger with a different selectivity compared with traditional ion exchangers. It can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges.

The small column volume of 4.7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.

If necessary, two columns can easily be connected in series to give a bed height of 20 cm.

Specifications
  • Particle Size:
    75 µm
  • Capacity:
    4.7 ml
  • Description:
    HiScreen™ Capto™ MMC
  • Bed dimensions:
    7.7×100 mm
  • Cat. No.:
    CA97067-800
  • Max. pressure:
    3 bar [0.3 MPa] (44 psi)
  • Storage:
    4...30 °C, 20% Ethanol
  • pH range:
    2 - 12
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.