PlasmidSelect Xtra Hydrophobic Interaction Chromatography Media, Cytiva
Catalog # CA97067-766
Supplier: Cytiva
CAS Number:
Specifications
- Particle Size:34 µm
- Description:PlasmidSelect Xtra
- Size:200 mL
- pH range:3 - 11
- Matrix:Cross-linked agarose
- Ligand:2-mercaptopyridine
- Storage temperature:4 - 30 °C
- Flow rate:<120 cm/h
- Cat. no.:CA97067-766
- Type:Liquid
- Supplier No.:28402402
Specifications
About this item
PlasmidSelect Xtra is a thiophilic aromatic adsorption chromatography medium with a selectivity that allows generic purification of supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
- Allows for generic purification of supercoiled plasmid DNA
- Allows for consistent purification results when scaling up from research scale to cGMP production
- Hydrophilic nature of the base matrix ensures low levels of non specific binding
- BioProcess medium supported for industrial applications
Available in convenient prepacked format for research, process development and quantitative and qualitative analysis of supercoiled plasmid DNA.
Degree of substitution: Approx. 3.5 mg 2-mercaptopyridine/ml medium
Storage conditions: 4 to 30 °C, 20% Ethanol
Chemical stability: Stable in commonly used aqueous buffers