Order Entry
Canada
ContactUsLinkComponent
Capto™ Q ImpRes Ion Exchange Chromatography Media, Cytiva
Capto™ Q ImpRes Ion Exchange Chromatography Media, Cytiva
Supplier:  Cytiva
Certificates
undefined
Capto™ Q ImpRes Ion Exchange Chromatography Media, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    2 - 12
  • Matrix:
    High-flow agarose
  • Ligand:
    Quaternary amine
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    >55 mg BSA, >48 mg β-lactoglobulin
  • Total ionic capacity:
    0,15 - 0,18 mmol Cl-/ml

Specifications

Frequently Bought Together