Order Entry
Canada
ContactUsLinkComponent
 
SOURCE™ 15Q Ion Exchange Chromatography Media, Cytiva
 :  Cytiva
 
undefined
SOURCE™ 15Q Ion Exchange Chromatography Media, Cytiva
 :  Cytiva
 

 

  • pH range:
    2 - 12
  • Matrix:
    Polystyrene/divinylbenzene
  • Ligand:
    Quaternary ammonium
  • Storage temperature:
    4 - 30 °C
  • Ligand coupling method:
    Long, hydrophilic spacer arms

 

Frequently Bought Together