HiLoad Superdex Columns, 30 PG, Cytiva
Supplier: Cytiva
Certificates
About this item
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component.
- High resolution with short run times and good recovery
- High capacity at high flow rates
- Separations can be scaled up to production levels
- Prepacked for convenience and reproducibility
- ¹/₁₆" fittings for convenient connections to ÄKTA design systems
Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr <10000 (Superdex 30 pg), Mr 3000 to 70000 (Superdex 75 pg), and Mr 10000 600000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.
Delivery: Includes dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs and instructions.
Specifications
- Catalog No:
- CA71002-662
- CA71002-664
- Max. pressure:
- 3 bar [0.3 MPa] (42 psi)
- 3 bar [0.3 MPa] (42 psi)
- pH range:
- 3 - 12
- 3 - 12
- Storage:
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol
Specifications
Related Information
Certificates
Search by Lot Number28989331_HAND BOOK 1.pdf 28989331_HAND BOOK.pdf 28989331_INSTRUCTIONS 1.pdf 28989331_INSTRUCTIONS.pdf 28989332_HAND BOOK 1.pdf
Manual
More
CyDye Licensing Statements GE Healthcare Life Sciences Cytiva CyDye Licensing Statements Cytiva General Licensing Statements Licensing Statements GE Healthcare Life Sciences
Additional Product Details
GE Healthcare SEC Columns and Media Selection Guide
Product Literature
Frequently Bought Together