Order Entry
Canada
ContactUsLinkComponent
nProtein A Sepharose™ 4 Fast Flow Affinity Chromatography Media, Cytiva
nProtein A Sepharose™ 4 Fast Flow Affinity Chromatography Media, Cytiva
Supplier:  Cytiva
Certificates
undefined
nProtein A Sepharose™ 4 Fast Flow Affinity Chromatography Media, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    3 - 9
  • Matrix:
    4% cross-linked agarose
  • Ligand:
    Native Protein A (S. aureus)
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    > 35 mg human IgG/mL

Specifications

Frequently Bought Together