HiTrap Columns, Protein L Columns Prepacked with Capto L, Cytiva
Supplier: Cytiva
Certificates
About this item
Columns prepacked with Capto™ L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.
- Prepacked for easy screening and process development, as well as small scale purification
- Columns easily connected in series for increased capacity
- Designed for use with syringe, peristalic pump, or chromatography system
Specifications
- Catalog No:
- CA11033-022
- CA11011-234
- CA11011-236
- CA11011-238
- Max. pressure:
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- 5 bar [0.5 MPa] (70 psi)
- pH range:
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- Storage:
- 2...8 °C, 20% Ethanol
- 2 to 8 °C, 20% Ethanol
- 2 to 8 °C, 20% Ethanol
- 2 to 8 °C, 20% Ethanol
Specifications
Related Information
Frequently Bought Together